Order Entry
Germany
Orders LinkContactUsLinkComponent
 

cis-4-Hydroxy-L-(-)-proline

Glass bottle for solids, 5 g
Catalog # BACE4011503.0005
Marketsource
Supplier:   BACHEM BIOCHEMICA
cis-4-Hydroxy-L-(-)-proline
Catalog # BACE4011503.0005
Supplier:  BACHEM BIOCHEMICA
Marketsource
CAS Number:  618-27-9
cis-4-Hydroxy-L-(-)-proline

Glass bottle for solids, 5 g

CAS Number:  618-27-9
Price
Inquire For Price
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  L(-)-cis-4-Hydroxyproline, H-cis-Hyp-OH, (2S,4S)-4-Hydroxypyrrolidine-2-carboxylic acid, (S)-(-)-cis-4-Hydroxyproline
  • CAS Number:618-27-9
  • Formula:C₅H₉NO₃
  • MDL Number:MFCD00064319
  • Molecular Weight:131.13 g/mol
  • Melting Pt:257 °C
  • EINECS:210-542-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications