D-(-)-Sorbitol ≥96%, high purity
Bucket, 5 kg
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
- CAS Number:50-70-4
- Formula:HOH₂C(CH(OH))₄CH₂OH
- MDL Number:MFCD00004708
- Molecular Weight:182.17 g/mol
- Merck Index:13,08797
- Boiling Pt:295 °C (1013 hPa)
- Melting Pt:98…100 °C
- Density:1.524 g/cm³ (20 °C)
- Flash Pt:>100 °C
- Storage Temperature:Ambient
- EINECS:200-061-5
Documents
Specifications
Specification Test Results
- Arsenic:<0.0003%
- Chloride:<0.005%
- Heavy Metals:<0.001%
- Moisture (KF):≤1%
- Purity (Anhydrous):≥96%
- Residue after Ignition:<0.1%
- Sulphate:<0.01%
Specifications
- Pack type:Bucket
- Pk:5 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



