Tween® 80 (Polysorbate), Proteomics Grade
HDPE bottle, 1 L
Supplier: VWR Chemicals
Tween® 80 (Polysorbate), Proteomics Grade
Catalog # 0442-1L
Supplier: VWR Chemicals
CAS Number: 9005-65-6
Jump to:
Product Details & Documents
Non-ionic detergent efficiently separates hydrophilic proteins from membrane spanning, hydrophobic proteins without altering biological activity.
- CAS Number:9005-65-6
- MDL Number:MFCD00082107
- Merck Index:13,07664
- Boiling Pt:>100 °C (1013 hPa)
- Melting Pt:–21 °C
- Density:1.062 g/cm³ (20 °C)
- Flash Pt:149 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:500-019-9
Documents
Specifications
Specification Test Results
- DNase:None
- Heavy metals:<0.001%
- Hydroxyl number:65 − 80
- Identification:Pass
- Moisture (KF):3.0%
- Protease:None
- Residue on ignition:0.25%
- RNase:None
- Specific gravity:1.06 − 1.09
- Viscosity:300 − 500 centistokes
Specifications
- Pack type:HDPE bottle
- Pk:1 L



