Orders LinkContactUsLinkComponent
Maintenance sorbents
Maintenance sorbents
Minipillows, 32 litres/box, Box of 16 minipillows, 380×180 mm
Catalog # 129-0146
Supplier:   3M
Maintenance sorbents
Catalog # 129-0146
Supplier:  3M
Maintenance sorbents
Maintenance sorbents
Minipillows, 32 litres/box, Box of 16 minipillows, 380×180 mm
Price
133,00 €
Log in for availability
1 * 16 ST
Product Details & Documents
Light grey when dry and darken when wet as an indication of saturation level.
  • Inert, synthetic fibres principally of PP and polyester
  • Multi-format combines four formats in one product and can be used as booms, pillows, sheets or rolls
  • Minibooms are designed for placing around the base of equipment to contain and control leaks and spills
  • Minipillows are high capacity for use at the source of a leak. They are tear resistant and can be left in place for long periods
Used for the control of accidental and routine leaks, drips and spills of oils, lubricants and other non aggressive liquids. They are perfect for cleaning up non aggressive fluids as well as being an effective maintenance management tool.
Specifications
  • Description:
    Minipillows
  • L×W:
    380×180 mm
  • Absorbency:
    32 litres/box
  • Packed:
    Box of 16 minipillows