Screw caps with aperture
GL 18, Screw cap with hole Ø 11,0 mm
Catalog # 201-0247
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
PBT, red without seals.
- For DIN GL threads
- Heat resistant up to 180 °C
- Autoclavable, sterilisable with hot air
With central hole.
Specifications
- Thread:GL 18
- Description:Screw cap with hole Ø 11,0 mm
- Seq:02
Accessories
Product Information
- Catalog #ForAvailabilityPrice
Product Information
- Catalog #ForAvailabilityPrice
Product Information
- Catalog #ForAvailabilityPrice
Product Information
- Catalog #ForAvailabilityPrice



