Order Entry
Germany
Orders LinkContactUsLinkComponent
Methylcyclohexane, Supelco®

Methylcyclohexane, Supelco®

Glass bottle, 5 mL
Catalog # 66294-5ML
Flammable
Health
Irritant
Environment
Supplier:   Merck
Methylcyclohexane, Supelco®
Catalog # 66294-5ML
Supplier:  Merck
CAS Number:  108-87-2
Methylcyclohexane, Supelco®
Methylcyclohexane, Supelco®

Glass bottle, 5 mL

CAS Number:  108-87-2
Price
247,00 €
Log in for availability
1 * 5 mL
Product Details & Documents
  • CAS Number:108-87-2
  • Formula:C₆H₁₁CH₃
  • MDL Number:MFCD00001497
  • Molecular Weight:98.19 g/mol
  • Merck Index:13,06074
  • Boiling Pt:101 °C (1013 hPa)
  • Melting Pt:–126 °C
  • Density:0.7693 g/cm³ (20 °C)
  • Flash Pt:–4 °C
  • Storage Temperature:Ambient
  • UN:2296
  • ADR:3,II
  •  :M-Clarity™ quality level = MQ100
  • EINECS:203-624-3

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.