Order Entry
Germany
Orders LinkContactUsLinkComponent
Sub-surface samplers, Wheaton® Grab Sampler® II

Sub-surface samplers, Wheaton® Grab Sampler® II

Grab Sampler® II with sample bottle and PTFE-lined cap, 1000 ml, 183 cm
Catalog # WHEA990350
Supplier:   DWK Life Sciences
Sub-surface samplers, Wheaton® Grab Sampler® II
Catalog # WHEA990350
Supplier:  DWK Life Sciences
Sub-surface samplers, Wheaton® Grab Sampler® II
Sub-surface samplers, Wheaton® Grab Sampler® II

Grab Sampler® II with sample bottle and PTFE-lined cap, 1000 ml, 183 cm

Price
1 620,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

The high density PP head holds a narrow mouth borosilicate jar.

  • Remote valve that can be opened by pulling and releasing the finger ring on the handle
  • Aluminum sections can be broken down for easy transport
  • Three lengths available

Certifications: EPA requirements for water and waste water sampling.

Specifications
  • Capacity:
    1000 ml
  • Description:
    Grab Sampler® II with sample bottle and PTFE-lined cap
  • Length:
    183 cm
  • Seq:
    01
Accessories

Product Information

  • Catalog #
    Capacity
    Thread
    Availability
    Price
  • WHEA990477
    1000 ml
    38-430
    Log in for availability
    1 * 1 ST - 98,50 €

Product Information

  • Catalog #
    Thread
    Colour
    Availability
    Price
  • Screw cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber liner
    Screw cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber liner
    WHEA240481
    38-430
    Black
    Log in for availability
    1 * 100 ST - 631,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.