Drum thieves, Wheaton®
Drum thieves, Glass, 150 ml
Catalog # WHEA885303
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
Drum thieves made of soda lime glass for quick disposal.
- Conventional open end drum thieves
- Just insert empty tube in drum
- Hold thumb over top and withdraw
Specifications
- Capacity:150 ml
- Description:Drum thieves
- Material:Glass