Orders LinkContactUsLinkComponent
Drum thieves, Wheaton®
Drum thieves, Wheaton®
Drum thieves, Glass, 75 ml
Catalog # WHEA885302
Supplier:   DWK Life Sciences
Drum thieves, Wheaton®
Catalog # WHEA885302
Supplier:  DWK Life Sciences
Drum thieves, Wheaton®
Drum thieves, Wheaton®
Drum thieves, Glass, 75 ml
Price
200,00 €
Log in for availability
1 * 24 ST
Product Details & Documents
Drum thieves made of soda lime glass for quick disposal.
  • Conventional open end drum thieves
  • Just insert empty tube in drum
  • Hold thumb over top and withdraw
Specifications
  • Capacity:
    75 ml
  • Description:
    Drum thieves
  • Material:
    Glass