Orders LinkContactUsLinkComponent
BOD bottles, WHEATON®
BOD bottles, WHEATON®
BOD bottles, with bar codes, borosilicate glass, 69×165 mm (Ø×H), number sequence: 241-264, 300 ml
Artikelnummer WHEA227497-11
Hersteller:   DWK Life Sciences
BOD bottles, WHEATON®
Artikelnummer WHEA227497-11
Hersteller:  DWK Life Sciences
BOD bottles, WHEATON®
BOD bottles, WHEATON®
BOD bottles, with bar codes, borosilicate glass, 69×165 mm (Ø×H), number sequence: 241-264, 300 ml
Preis
787,00 €
Log in for availability
1 * 24 ST
Product Details & Documents
Made of Type I borosilicate glass.
  • Available with or without stopper
  • With white labeling area
Specifications
  • Capacity:
    300 ml
  • Description:
    BOD bottles, with bar codes, borosilicate glass, 69×165 mm (Ø×H), number sequence: 241-264