Order Entry
Germany
Orders LinkContactUsLinkComponent
BOD bottles, WHEATON®

BOD bottles, WHEATON®

BOD bottles, borosilicate glass, 43×115 mm (Ø×H), unnumbered, 60 ml
Catalog # WHEA227494-00G
Supplier:   DWK Life Sciences
BOD bottles, WHEATON®
Catalog # WHEA227494-00G
Supplier:  DWK Life Sciences
BOD bottles, WHEATON®
BOD bottles, WHEATON®

BOD bottles, borosilicate glass, 43×115 mm (Ø×H), unnumbered, 60 ml

Price
1 930,00 €
Log in for availability
1 * 36 ST
Product Details & Documents

Made of Type I borosilicate glass.

  • Available with or without stopper
  • With white labeling area
Specifications
  • Capacity:
    60 ml
  • Description:
    BOD bottles, borosilicate glass, 43×115 mm (Ø×H), unnumbered
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.