Dropping bottles, LDPE, WHEATON®
7 ml, LDPE, white, 15-425
Catalog # 215-4000
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
LDPE, translucent, with white PP cap.
- Good chemical resistance, heat resistant up to 80 °C
- Bottle dispenses one drop at a time
- Dropper tip snaps firmly in place and is easily removed for filling
With extended controlled dropper tip and dropper tip cap. These dropping bottles provide reliable, repeatable dispensing of reagents and other liquids. They serve as a very good alternative to pipettes or other conventional dispensing devices. They are suitable for most biotech, diagnostic and pharmaceutical applications.
Specifications
- Thread:15-425
- Capacity:7 ml
- Material:LDPE, white
- Seq:03



