Snap caps for sample bottles, WHEATON®
30 mm, 16/24/30/37/44 ml
Catalog # WHEA242615
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
Natural low density polyethylene.
- All sizes have tabs for easy removal
- Good chemical resistance
- Good temperature resistance
For WHEATON snap cap sample bottles.
Specifications
- Cap Size:30 mm
- For bottles:16/24/30/37/44 ml