Snap caps for sample bottles, WHEATON®
45 mm, 60/120 ml
WHEA242619
Natural low density polyethylene.
- All sizes have tabs for easy removal
- Good chemical resistance
- Good temperature resistance
For WHEATON snap cap sample bottles.
- Cap Size:45 mm
- For bottles:60/120 ml