Orders LinkContactUsLinkComponent
Screw caps, WHEATON®
Screw caps, WHEATON®
Screw cap, solid top, phenolic, with PE Poly-Seal® liner, 22-400, Black
Catalog # WHEA239255
Supplier:   DWK Life Sciences
Screw caps, WHEATON®
Catalog # WHEA239255
Supplier:  DWK Life Sciences
Screw caps, WHEATON®
Screw caps, WHEATON®
Screw cap, solid top, phenolic, with PE Poly-Seal® liner, 22-400, Black
Price
110,00 €
Log in for availability
1 * 144 ST
Product Details & Documents
Rubber lined caps feature a non-toxic white styrene-butadiene rubber liner ideal for cell culture work.
  • Aluminum foil faced pulp board liners can be used with organic solvents. Good resistance to hydrocarbons, oils, ketones and alcohols Not good for acids or alkalis.
Use low density polyethylene (LDPE) lined caps with distilled water, analytical standards and reagents.
PTFE faced (14B) styrene-butadiene rubber liner provides a totally inert inner seal and surface facing the sample or product.
Bonded PTFE faced silicone liner ideal for storage of volatile solvents, sensitive compounds and corrosive chemicals.
Poly-Seal liner (LDPE) has unique cone liner design and offers superior torque retention and excellent sealing characteristics.
Caution: It is recommended that liner be tested prior to use for leak resistant applications.
Specifications
  • Thread:
    22-400
  • Colour:
    Black
  • Description:
    Screw cap, solid top, phenolic, with PE Poly-Seal® liner
  • Seq:
    41