Orders LinkContactUsLinkComponent
Replacement impeller for Magna Flex™ flasks
Replacement impeller for Magna Flex™ flasks
Replacement impeller for 8000 ml Magna Flex® spinner flask
Catalog # 734-3149
Supplier:   DWK Life Sciences
Replacement impeller for Magna Flex™ flasks
Catalog # 734-3149
Supplier:  DWK Life Sciences
Replacement impeller for Magna Flex™ flasks
Replacement impeller for Magna Flex™ flasks
Replacement impeller for 8000 ml Magna Flex® spinner flask
Price
341,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
This unit includes a top cap with liners, glass stirring ball and connecting silicone tubing. This glass impeller assembly is designed for replacement on all Magna Flex™ style flasks.
Specifications
  • Description:
    Replacement impeller for 8000 ml Magna Flex® spinner flask
  • Seq:
    07