Order Entry
Germany
Orders LinkContactUsLinkComponent
Raschig rings

Raschig rings

Raschig ring, 4 mm, 4 mm
Catalog # LENZ05127004
Supplier:   Lenz Laborglas
Raschig rings
Catalog # LENZ05127004
Supplier:  Lenz Laborglas
Raschig rings
Raschig rings

Raschig ring, 4 mm, 4 mm

Price
141,00 €
Log in for availability
1 * 1 kg
Product Details & Documents

Soda lime glass.

  • For packing columns
  • Good chemical resistance
  • Good thermal resistance
Specifications
  • Ext.Ø:
    4 mm
  • Description:
    Raschig ring
  • Height:
    4 mm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.