Agarose
1 KIT
Catalog # TREV9700-025-P
Marketsource
Supplier: TREVIGEN
Jump to:
Product Details & Documents
- CAS Number:9012-36-6
- MDL Number:MFCD00081294
- Melting Pt:60…90 °C
- Storage Temperature:Ambient
- EINECS:232-731-8
Specifications
Specifications
- Pk:1 KIT



