Auftragserfassung
Germany
AuftragsverfolgungKontakt
Extraction Buffer 3 8 ml 1 * 8 mL

Extraction Buffer 3 8 ml 1 * 8 mL

Catalog # TREV4850-20-03
Marketsource
Supplier:   TREVIGEN
Extraction Buffer 3 8 ml 1 * 8 mL
Catalog # TREV4850-20-03
Supplier:  TREVIGEN
Marketsource
Extraction Buffer 3 8 ml 1 * 8 mL
Price
Inquire For Price
Log in for availability
1 * 8 mL
Product Details & Documents
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.