Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
FLARE Buffer 1, 25 x 40 ml 1 * 40 mL
Catalog # TREV3950-040-01
Supplier:
TREVIGEN
Expand image
FLARE Buffer 1, 25 x 40 ml 1 * 40 mL
Catalog # TREV3950-040-01
Supplier:
TREVIGEN