Filter papers, reusable, Aesculap®
95×215 mm
Catalog # AESCJK091
Supplier: Aesculap
Jump to:
Product Details & Documents
Reusable filters for 1000 cleaning and sterilisation cycles.
Specifications
- Seq:01
- Size:95×215 mm