Order Entry
Germany
Orders LinkContactUsLinkComponent
Gouges, Aesculap®

Gouges, Aesculap®

Gouge for scoliose operating, Zielke, 9 mm, 230 mm
Catalog # AESCFL702R
Supplier:   Aesculap
Gouges, Aesculap®
Catalog # AESCFL702R
Supplier:  Aesculap
Gouges, Aesculap®

Gouge for scoliose operating, Zielke, 9 mm, 230 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Type:
    Zielke
  • Width:
    9 mm
  • Description:
    Gouge for scoliose operating
  • Length:
    230 mm
  • Seq:
    40
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.