Order Entry
Germany
Orders LinkContactUsLinkComponent
Gouges, Aesculap®

Gouges, Aesculap®

Gouge for removing broken screws, Lexer, 9 mm, 220 mm
Catalog # AESCFL420R
Supplier:   Aesculap
Gouges, Aesculap®
Catalog # AESCFL420R
Supplier:  Aesculap
Gouges, Aesculap®

Gouge for removing broken screws, Lexer, 9 mm, 220 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Type:
    Lexer
  • Width:
    9 mm
  • Description:
    Gouge for removing broken screws
  • Length:
    220 mm
  • Seq:
    38