Order Entry
Germany
Orders LinkContactUsLinkComponent
Gouges, Aesculap®

Gouges, Aesculap®

Gouge, Lexer, 30 mm, 220 mm
Catalog # AESCFL418R
Supplier:   Aesculap
Gouges, Aesculap®
Catalog # AESCFL418R
Supplier:  Aesculap
Gouges, Aesculap®

Gouge, Lexer, 30 mm, 220 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Type:
    Lexer
  • Width:
    30 mm
  • Description:
    Gouge
  • Length:
    220 mm
  • Seq:
    15
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.