Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Gouges, Aesculap®

Gouge, Alexander, 11 mm, 170 mm
  AESCFL347R
 :   Aesculap
Gouges, Aesculap®
  AESCFL347R
 :  Aesculap
Gouges, Aesculap®

Gouge, Alexander, 11 mm, 170 mm

 
 
   
1 * 1 ST
 
 
  • Type:
    Alexander
  • Width:
    11 mm
  • Description:
    Gouge
  • Length:
    170 mm
  • Seq:
    08
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.