Order Entry
Germany
Orders LinkContactUsLinkComponent
Gouges, Aesculap®

Gouges, Aesculap®

Gouge, Partsch, 4 mm, 140 mm
Catalog # AESCFL214R
Supplier:   Aesculap
Gouges, Aesculap®
Catalog # AESCFL214R
Supplier:  Aesculap
Gouges, Aesculap®

Gouge, Partsch, 4 mm, 140 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Type:
    Partsch
  • Width:
    4 mm
  • Description:
    Gouge
  • Length:
    140 mm
  • Seq:
    26