Order Entry
Germany
Orders LinkContactUsLinkComponent
Gouges, Aesculap®

Gouges, Aesculap®

Gouge, Mannerfelt, 3 mm, 155 mm
Catalog # AESCFL153R
Supplier:   Aesculap
Gouges, Aesculap®
Catalog # AESCFL153R
Supplier:  Aesculap
Gouges, Aesculap®

Gouge, Mannerfelt, 3 mm, 155 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Type:
    Mannerfelt
  • Width:
    3 mm
  • Description:
    Gouge
  • Length:
    155 mm
  • Seq:
    23
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.