Order Entry
Germany
Orders LinkContactUsLinkComponent
Bone curettes, Aesculap®

Bone curettes, Aesculap®

Bone curette, Volkmann type (Fig. 6), with 14,5 mm end, 170 mm
Catalog # AESCFK639R
Supplier:   Aesculap
Bone curettes, Aesculap®
Catalog # AESCFK639R
Supplier:  Aesculap
Bone curettes, Aesculap®

Bone curette, Volkmann type (Fig. 6), with 14,5 mm end, 170 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Bone curette, Volkmann type (Fig. 6), with 14,5 mm end
  • Length:
    170 mm