Order Entry
Germany
Orders LinkContactUsLinkComponent
Bone curettes, Aesculap®

Bone curettes, Aesculap®

Bone curette, Volkmann type (Fig. 5), with 13 mm end, 170 mm
Catalog # AESCFK638R
Supplier:   Aesculap
Bone curettes, Aesculap®
Catalog # AESCFK638R
Supplier:  Aesculap
Bone curettes, Aesculap®

Bone curette, Volkmann type (Fig. 5), with 13 mm end, 170 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Bone curette, Volkmann type (Fig. 5), with 13 mm end
  • Length:
    170 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.