Order Entry
Germany
Orders LinkContactUsLinkComponent
Dilator set, Aesculap®

Dilator set, Aesculap®

Dilator, 25 mm
Catalog # AESCEM125R
Supplier:   Aesculap
Dilator set, Aesculap®
Catalog # AESCEM125R
Supplier:  Aesculap
Dilator set, Aesculap®

Dilator, 25 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

Complete set of uterine dilators.

Delivery information: Consists of 26 dilators, 1 rack for dilators (1 to 17 mm), 1 rack for dilators (18 to 26 mm) and metal case with hinged aluminium lid (330×220×80 mm).

Specifications
  • Description:
    Dilator, 25 mm
  • Seq:
    26