Order Entry
Germany
Orders LinkContactUsLinkComponent
Dilator set, Aesculap®

Dilator set, Aesculap®

Dilator, 3 mm
Catalog # AESCEM103R
Supplier:   Aesculap
Dilator set, Aesculap®
Catalog # AESCEM103R
Supplier:  Aesculap
Dilator set, Aesculap®

Dilator, 3 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

Complete set of uterine dilators.

Delivery information: Consists of 26 dilators, 1 rack for dilators (1 to 17 mm), 1 rack for dilators (18 to 26 mm) and metal case with hinged aluminium lid (330×220×80 mm).

Specifications
  • Description:
    Dilator, 3 mm
  • Seq:
    04
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.