Orders LinkContactUsLinkComponent
Splinter forceps, Aesculap®
Splinter and dissecting forceps, Curved, serrated, very fine tips, 130 mm
Catalog # AESCBD314R
Supplier:   Aesculap
Splinter forceps, Aesculap®
Catalog # AESCBD314R
Supplier:  Aesculap
Splinter forceps, Aesculap®
Splinter and dissecting forceps, Curved, serrated, very fine tips, 130 mm
Price
25,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Type:
    Curved, serrated, very fine tips
  • Description:
    Splinter and dissecting forceps
  • Length:
    130 mm
  • Seq:
    09