Dissecting forceps, Aesculap®
Dissecting forceps, Gerald, Curved, serrated, very fine tips, 175 mm
Catalog # AESCBD229R
Supplier: Aesculap
Jump to:
Product Details & Documents
Stainless steel.
- Very good chemical resistance
- Very good temperature resistance
- Easy to clean
Delicate dissecting forceps with plateau and many types.
Specifications
- Type:Curved, serrated, very fine tips
- Description:Dissecting forceps, Gerald
- Length:175 mm
- Seq:10