Orders LinkContactUsLinkComponent
Dissecting forceps, Aesculap®
Dissecting forceps, Aesculap®
Dissecting forceps, SEMKEN, Straight, serrated, very fine tips, 155 mm
Catalog # AESCBD217R
Supplier:   Aesculap
Dissecting forceps, Aesculap®
Catalog # AESCBD217R
Supplier:  Aesculap
Dissecting forceps, Aesculap®
Dissecting forceps, SEMKEN, Straight, serrated, very fine tips, 155 mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel.
  • Very good chemical resistance
  • Very good temperature resistance
  • Easy to clean
Delicate dissecting forceps with plateau and many types.
Specifications
  • Type:
    Straight, serrated, very fine tips
  • Description:
    Dissecting forceps, SEMKEN
  • Length:
    155 mm
  • Seq:
    09