Dissecting forceps, Aesculap®
Finochietto, curved, with eye, 245 mm
Catalog # AESCBJ052R
Supplier: Aesculap
Jump to:
Product Details & Documents
Specifications
- Type:Finochietto, curved, with eye
- Length:245 mm
- Seq:10