Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SOLVENT FLOW APPARATUS W/ 500ML VESSEL 1 * 1 items
Catalog # QVFLLSG0880103
Supplier:
QVF LABORTECHNIK
Expand image
SOLVENT FLOW APPARATUS W/ 500ML VESSEL 1 * 1 items
Catalog # QVFLLSG0880103
Supplier:
QVF LABORTECHNIK