Order Entry
Germany
Orders LinkContactUsLinkComponent
Weighing scoops

Weighing scoops

160 mm, 30 - 75 mm
Catalog # 611-9715
Supplier:   BOCHEM
Weighing scoops
Catalog # 611-9715
Supplier:  BOCHEM
Weighing scoops
Weighing scoops

160 mm, 30 - 75 mm

Price
36,20 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel 18/10, anti-magnetic.

Specifications
  • Width:
    30 - 75 mm
  • Length:
    160 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.