Order Entry
Germany
Orders LinkContactUsLinkComponent
Beakers, low form

Beakers, low form

100 ml, 60 mm, 50 mm
Catalog # BOCH8510
Supplier:   BOCHEM
Beakers, low form
Catalog # BOCH8510
Supplier:  BOCHEM
Beakers, low form
Beakers, low form

100 ml, 60 mm, 50 mm

Price
80,50 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel, 18/10.

  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning

Ideal for corrosive materials. Robust and durable.

Specifications
  • Ext.Ø:
    50 mm
  • Capacity:
    100 ml
  • Height:
    60 mm