Sulphite standard solution
Sulfite standard solution, pack of 16×10 ml voluette ampoules (NIST), 5000 mg/l as SO3, 16
Catalog # HACH2267410
Supplier: Hach
Jump to:
Product Details & Documents
This equivalent standard is prepared with stabilised sodium thiosulfate, since sulphite is not a stable in solution.
Certificates
Search by Lot NumberSpecifications
Specifications
- Concentration:5000 mg/l as SO<sub>3</sub>
- Pk:16
- Description:Sulfite standard solution, pack of 16×10 ml voluette ampoules (NIST)