Order Entry
Germany
Orders LinkContactUsLinkComponent
Diethyl ether stabilised, TECHNICAL, rectified

Diethyl ether stabilised, TECHNICAL, rectified

Aluminium bottle, 5 L
Catalog # 23807.366
Flammable
Irritant
Supplier:   VWR Chemicals
Diethyl ether stabilised, TECHNICAL, rectified
Catalog # 23807.366
Supplier:  VWR Chemicals
CAS Number:  60-29-7
Diethyl ether stabilised, TECHNICAL, rectified
Diethyl ether stabilised, TECHNICAL, rectified

Aluminium bottle, 5 L

CAS Number:  60-29-7
Price
134,00 €
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
  • CAS Number:60-29-7
  • Formula:(CH₃CH₂)₂O
  • MDL Number:MFCD00011646
  • Molecular Weight:74.12 g/mol
  • Merck Index:12,03852
  • Boiling Pt:35 °C (1013 hPa)
  • Melting Pt:–123 °C
  • Density:0.7135 g/cm³ (20 °C)
  • Flash Pt:–40 °C
  • UN:1155
  • ADR:3,I
  •  :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 4-10 ppm
  • Storage Temperature:Ambient
  • EINECS:200-467-2
  • REACH:01-2119535785-29
Specifications

Specification Test Results

  • Identification:
    Passes test

Specifications

  • Pack type:
    Aluminium bottle
  • Pk:
    5 L
  • Cat. No.:
    23807.366

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.