Order Entry
Germany
Orders LinkContactUsLinkComponent
Acetone ≥99%, TECHNICAL

Acetone ≥99%, TECHNICAL

Plastic container, 5 L
Catalog # 20063.365
Flammable
Irritant
   Safer Human and Environmental Health
Supplier:   VWR Chemicals
Acetone ≥99%, TECHNICAL
Catalog # 20063.365
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  67-64-1
Acetone ≥99%, TECHNICAL
Acetone ≥99%, TECHNICAL

Plastic container, 5 L

CAS Number:  67-64-1
Price
72,50 €
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Merck Index:13,00067
  • Boiling Pt:56.2 °C (1013 hPa)
  • Melting Pt:–95.4 °C
  • Density:0.792 g/cm³ (20 °C)
  • Flash Pt:<–20 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-662-2
  • UN:1090
  • ADR:3,II
  • REACH:01-2119471330-49
Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • IR Spectrum:
    Passes test

Specifications

  • Pack type:
    Plastic container
  • Pk:
    5 L
  • Cat. No.:
    20063.365
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.