Robotic tips for Beckman®
919-262-05 (Low Retention), 250 µl, -, -, Transparent, 10 racks, 96 each
Catalog # 736-0100
Supplier: Thermo Fisher Scientific
Robotic tips for Beckman®
Catalog # 736-0100
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
High quality tips, tested on the workstations for which they were created, to ensure compatibility and a secure fit. Certified free of RNase, DNase, DNA, RNA, pyrogens and ATP.
- Transparent PP
- Tips with ART® filters: Eliminate any risk of cross-contamination between samples; designed for applications with DNA, RNA as well as radioactive and infectious samples
- Low retention tips: Hydrophobic tips retain less sample which decreases the amount of reagent loss while significantly increasing result reproducibility
- Sterile tips: Sterilised by gamma radiation
Caution: * Requires adapter 3772 only available directly from Molecular BioProducts.
Documents
Certificates
Search by Lot NumberSpecifications
- Packed:10 racks, 96 each
- Sterile:-
- Volume:250 µl
- Tip style:919-262-05 (Low Retention)
- Tip colour:Transparent
- Filter:-
- Cat. No.:736-0100
- Environmental certification:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Low Manufacturing Impact
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



