Orders LinkContactUsLinkComponent
Robotic tips for Beckman®
Robotic tips for Beckman®
918-011, 20 µl, +, +, Black, 10 racks, 96 each
Catalog # 736-0018
   Low Manufacturing Impact
Robotic tips for Beckman®
Catalog # 736-0018
   Low Manufacturing Impact
Robotic tips for Beckman®
Robotic tips for Beckman®
918-011, 20 µl, +, +, Black, 10 racks, 96 each
Price
365,00 €
Log in for availability
1 * 960 ST
Product Details & Documents
High quality tips, tested on the workstations for which they were created, to ensure compatibility and a secure fit. Certified free of RNase, DNase, DNA, RNA, pyrogens and ATP.
  • Transparent PP
  • Tips with ART® filters: Eliminate any risk of cross-contamination between samples; designed for applications with DNA, RNA as well as radioactive and infectious samples
  • Low retention tips: Hydrophobic tips retain less sample which decreases the amount of reagent loss while significantly increasing result reproducibility
  • Sterile tips: Sterilised by gamma radiation
Achtung! Bitte beachten: * Requires adapter 3772 only available directly from Molecular BioProducts.
Documents
Specifications
  • Packed:
    10 racks, 96 each
  • Sterile:
    +
  • Volume:
    20 µl
  • Tip style:
    918-011
  • Tip colour:
    Black
  • Filter:
    +
  • Cat. No.:
    736-0018
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact