SODIUM METHOXIDE, 0.5M SOLUTION IN METHA NOL, A.C.S. REAGENT 1 * 100 mL
Catalog # SIAL403067-100ML
Supplier: Merck
SODIUM METHOXIDE, 0.5M SOLUTION IN METHA NOL, A.C.S. REAGENT 1 * 100 mL
Catalog # SIAL403067-100ML
Supplier: Merck
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Neutralising agent for the automated cleaning of glassware, neodisher® Z
Supplier: Dr. Weigert
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Transfer pipettes
Supplier: RATIOLAB
Detergents, liquid, for automated cleaning of glassware, neodisher® FLA
Supplier: Dr. Weigert
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Heat resistant gloves, ActivArmr® 43-113
Supplier: Ansell