Orders LinkContactUsLinkComponent
Pasteur pipettes
Pasteur pipettes
Pasteur pipette, with open tip, 350 mm, -, 4× 250/box
Catalog # 612-0896
Supplier:   HILGENBERG
Pasteur pipettes
Catalog # 612-0896
Supplier:  HILGENBERG
Pasteur pipettes
Pasteur pipettes
Pasteur pipette, with open tip, 350 mm, -, 4× 250/box
Price
4 440,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
Pipettes made from soda lime glass.
  • With constriction
  • Available with open tip (unplugged) or with closed tip (with cotton plug)
Specifications
  • Packed:
    4× 250/box
  • Sterile:
    -
  • Description:
    Pasteur pipette, with open tip
  • Length:
    350 mm
  • Seq:
    02