Orders LinkContactUsLinkComponent
Polyethylene stoppers | Avantor
Polyethylene stoppers
White, PE, 14/23
Catalog # 217-0718
Supplier:   SciLabware
Polyethylene stoppers
Catalog # 217-0718
Supplier:  SciLabware
Polyethylene stoppers
Polyethylene stoppers
White, PE, 14/23
Price
29,70 €
Log in for availability
1 * 10 ST
Product Details & Documents
These PE stoppers are ideally suited for use with glass or plastic volumetric flasks and measuring cylinders etc. which have longer 'B' length sockets.
Large head provides a good grip and gives protection if glassware is accidently knocked over.
 : 10/package
Specifications
  • NS:
    14/23
  • Colour:
    White
  • Material:
    PE