Order Entry
Germany
Orders LinkContactUsLinkComponent
Process pH combination electrodes

Process pH combination electrodes

pH combination electrode, with sphere membrane, H 6381, 1 - 14, 0...+135 °C, Glass, KCl 3 mol/l, Ceramic, 12×320 mm, Screw plug head
Catalog # SCOI285110195
Supplier:   SI Analytics
Process pH combination electrodes
Catalog # SCOI285110195
Supplier:  SI Analytics
Process pH combination electrodes
Process pH combination electrodes

pH combination electrode, with sphere membrane, H 6381, 1 - 14, 0...+135 °C, Glass, KCl 3 mol/l, Ceramic, 12×320 mm, Screw plug head

Price
359,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Connection:
    Screw plug head
  • Ø×L:
    12×320 mm
  • Type:
    H 6381
  • Description:
    pH combination electrode, with sphere membrane
  • Material:
    Glass
  • Seq:
    13
  • Diaphr.:
    Ceramic
  • Electrolyte:
    KCl 3 mol/l
  • Temp.:
    0...+135 °C
  • pH:
    1 - 14