Order Entry
Germany
Orders LinkContactUsLinkComponent
Silica standard solution

Silica standard solution

Silica standard solution, 25 mg/l as SiO2 (NIST), 236 mL
Catalog # HACH2122531
Supplier:   Hach
Silica standard solution
Catalog # HACH2122531
Supplier:  Hach
CAS Number:  
Silica standard solution

Silica standard solution, 25 mg/l as SiO2 (NIST), 236 mL

CAS Number:  
Price
68,50 €
Log in for availability
1 * 236 mL
Product Details & Documents

Standard solutions for accuracy checks in silica determinations.

Specifications

Specifications

  • Concentration:
    25 mg/l as SiO<sub>2 </sub> (NIST)
  • Pk:
    236 mL
  • Description:
    Silica standard solution

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.