Order Entry
Germany
Orders LinkContactUsLinkComponent
QUANTOFIX® Test Strips, Arsenic

QUANTOFIX® Test Strips, Arsenic

QUANTOFIX® ammonium, 0 • 10 • 25 • 50 • 100 • 200 • 400 - mg/L - NH₄⁺; 10–350 - mg/L - NH₄⁺, Color reaction based on Neßler’s reagent, 170×125×48 mm (packaging), 100 Tests, 100 Tests
Catalog # 705-0316
Supplier:   MACHEREY-NAGEL
QUANTOFIX® Test Strips, Arsenic
Catalog # 705-0316
Supplier:  MACHEREY-NAGEL
CAS Number:  
QUANTOFIX® Test Strips, Arsenic
QUANTOFIX® Test Strips, Arsenic

QUANTOFIX® ammonium, 0 • 10 • 25 • 50 • 100 • 200 • 400 - mg/L - NH₄⁺; 10–350 - mg/L - NH₄⁺, Color reaction based on Neßler’s reagent, 170×125×48 mm (packaging), 100 Tests, 100 Tests

CAS Number:  
Price
57,50 €
Log in for availability
1 * 100 Tests
Product Details & Documents

QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. The test strips are intended for single use, maintenance or calibration are not required.

  • Results within seconds: Just dip, wait and read against multi-stage colour scale Always ready to use: No accessories necessary for evaluation Safe results: Desiccant in the stopper for optimal protection of the strips against humidity

The test strips allow the quick and easy determination of arsenic in solutions. They come with all necessary reagents so that the measurement can immediately be started. Within 20 to 30 minutes one gets reliable results.

Specifications

Specifications

  • Chemical method:
    Color reaction based on Neßler’s reagent
  • Pk:
    100 Tests
  • Description:
    QUANTOFIX® ammonium
  • Dimensions:
    170×125×48 mm (packaging)
  • Tests per kit:
    100 Tests
  • Range:
    0 • 10 • 25 • 50 • 100 • 200 • 400 - mg/L - NH₄⁺; 10–350 - mg/L - NH₄⁺
  • Cat. No.:
    705-0316
  • Parameters:
    Requires additional reagent
  • Contents:
    100 test strips in a tube, additional regaents, instruction leaflet in a box
  • Platform:
    Semi-quantitative test strips
  • Compliance:
    Hazardous material
  • Instrument:
    QUANTOFIX Relax
  • Shelf life (months):
    30 (from production)
  • Method:
    Ammonium - Nessler (bright yellow → orange)
  • Weight:
    230 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.