Order Entry
Germany
Orders LinkContactUsLinkComponent
 

4-Chlorophenol for synthesis

25 kg
  8.02254.9025
 
 
 :   Merck
4-Chlorophenol for synthesis
  8.02254.9025
 :  Merck
 :  106-48-9
4-Chlorophenol for synthesis

25 kg

 :  106-48-9
 
 
   
1 * 25 kg
 
 :  p-Chlorophenol
  •  :106-48-9
  •  :C₆H₅ClO
  •  :MFCD00002318
  •  :128.56 g/mol
  •  :13,02173
  •  :220 °C (1013 hPa)
  •  :41…44 °C
  •  :1.31 g/cm³ (20 °C)
  •  :121 °C
  •  :Ambient
  •  :2020
  •  :6.1,III
  •  :M-Clarity™ quality level = MQ200
  •  :203-402-6
 

 

  • Solubility:
    27 g/l (20 °C)
  • Melting Point:
    41 - 44 °C
  • Molar Mass:
    128.55 g/mol
  • Boiling Point:
    220 °C (1013 hPa)
  • Vapor Pressure:
    0.15 hPa (20 °C)
  • Flash Point:
    121 °C
  • Density:
    1.26 g/cm3 (45 °C)
  • Synonyms:
    4-Chloro-1-hydroxybenzene
  • Hill Formula:
    C6H5ClO
  • Chemical Formula:
    4-(Cl)C6H4OH

 

  • Pk:
    25 kg